missing translation for 'onlineSavingsMsg'
Learn More

7Alpha-Hydroxy-4-cholesten-3-one-d7 (contains 2% d0), TRC

Product Code. 30398585
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398585 10 mg -
30396823 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398585 Supplier Toronto Research Chemicals Supplier No. TRCH82513310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27H37D7O2
Molecular Weight (g/mol) 407.68
Synonym (7α)-7-Hydroxycholest-4-en-3-one-d7, 7α-Hydroxycholest-4-en-3-one-d7
SMILES [H][C@@]12[C@H](O)CC3=CC(CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h15,17-18,21-25,29H,6-14,16H2,1-5H3/t18-,21-,22+,23+,24-,25+,26+,27-/m1/s1/i1D3,2D3,17D
Chemical Name or Material 7alpha-Hydroxy-4-cholesten-3-one-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.