missing translation for 'onlineSavingsMsg'
Learn More

7ACC2, MedChemExpress

Product Code. p-200100850
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481168

Brand: Medchem Express HYD0713

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 7ACC2 is a potent monocarboxylate transporter (MCT) inhibitor with an IC50 of 11 nM for inhibition of [14C]-lactate influx. 7ACC2 is also a potent inhibitor of mitochondrial pyruvate transport. 7ACC2 is an anticancer agent through inhibition of lactate flux.

7ACC2 is a potent monocarboxylate transporter (MCT) inhibitor with an IC50 of 11 nM for inhibition of [14C]-lactate influx. 7ACC2 is also a potent inhibitor of mitochondrial pyruvate transport. 7ACC2 is an anticancer agent through inhibition of lactate flux.

Specifications

Chemical Name or Material 7ACC2
CAS 1472624-85-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H15NO4
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (161.64 mM; Need ultrasonic)
SMILES CN(C1=CC=C(C=C(C(O)=O)C(O2)=O)C2=C1)CC3=CC=CC=C3
Molecular Weight (g/mol) 309.32
Formula Weight 309.32
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.