missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7ACC2, MedChemExpress
Description
7ACC2 is a potent monocarboxylate transporter (MCT) inhibitor with an IC50 of 11 nM for inhibition of [14C]-lactate influx. 7ACC2 is also a potent inhibitor of mitochondrial pyruvate transport. 7ACC2 is an anticancer agent through inhibition of lactate flux.
Specifications
Specifications
| Chemical Name or Material | 7ACC2 |
| CAS | 1472624-85-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H15NO4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50 mg/mL (161.64 mM; Need ultrasonic) |
| SMILES | CN(C1=CC=C(C=C(C(O)=O)C(O2)=O)C2=C1)CC3=CC=CC=C3 |
| Molecular Weight (g/mol) | 309.32 |
| Formula Weight | 309.32 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?