missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 7550 Single-Use Nitrile Gloves, Powder-Free

Product Code. 16283152
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16283152

Brand: SHOWA™ 7550XL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for working environments where gloves stain easily: chemicals, oils, greases, coloring agents... Silicon free and high tear resistance!

  • 100% nitrile. Slicon-free. Electrostatic dissipative in use (ESD). Low-modulus formulation to improve fit and reduce hand fatigue
  • Textured finish on fingertips to enhance grip
  • Second skin feel, softer texture and easy donning
  • Latex-free to avoid type I and type IV latex allergies

Specifications

Certifications/Compliance (Europe) EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011
AQL 1.5
Color Black
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Thickness (Metric) 0.10 mm
Finger Style Textured
Model 7550XL
Packaging Type dispenser box
Port Size (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.