missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 7505PF Single-Use Nitrile Gloves, Powder-Free
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
Description
- Protects from a wide array of chemical hazards while avoiding type I and type IV latex allergies
- Latex, powder-free
- Low-modulus formulation to improve fit and reduce hand fatigue
- Easy to don and remove
- Category III, EN374 approved.
Specifications
Specifications
| Certifications/Compliance (Europe) | EU Regulation 425/2016, EN 420, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011 |
| Description | AQL 1,0 |
| Color | Blue |
| Disposable | Single-use |
| Hand | Ambidextrous |
| Laboratory/Cleanroom Use | Laboratory |
| Latex Free | Yes |
| Material | Nitrile |
| Powdered | Powder Free |
| Size | 2X-Large |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction