missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 7505PF Single-Use Nitrile Gloves, Powder-Free

Product Code. 16283142
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16283142

Brand: SHOWA™ 7505PFXS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cost efficient protection!

  • Protects from a wide array of chemical hazards while avoiding type I and type IV latex allergies
  • Latex, powder-free
  • Low-modulus formulation to improve fit and reduce hand fatigue
  • Easy to don and remove
  • Category III, EN374 approved.

Specifications

Certifications/Compliance (Europe) EU Regulation 425/2016, EN 420, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011
Description AQL 1,0
Color Blue
Disposable Single-use
Hand Ambidextrous
Laboratory/Cleanroom Use Laboratory
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Sterility Non-sterile
Thickness (Metric) 0.10 mm
Finger Style textured finger-ends
Packaging Type dispenser box
Port Size (Metric) 24 cm
Tear Rating 6 N
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.