missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 740 Y-P

Product Code. 16451848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451848 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451848 Supplier MedChemExpress Supplier No. HYP0175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 740 Y-P (740YPDGFR; PDGFR 740Y-P) is a potent and cell-permeable PI3K activator. 740 Y-P readily binds GST fusion proteins containing both the N- and C- terminal SH2 domains of p85 but fails to bind GST alone.

Specifications

Purity 98.0%
CAS 1236188-16-1
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-Kinase/protease
Molecular Weight (g/mol) 3270.7 g/mol
Product Type Reference compound
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (7.64 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (0.61 mM∣Need ultrasonic)
Synonym 740YPDGFR∣ PDGFR 740Y-P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.