Learn More
Applied Biosystems™ 7300 System SDS v1.4 Software Upgrade Kit
Tous les produits Applied Biosystems ProduitsDescription
This software upgrades earlier versions of the 7300 SDS Software to provide maximum capability and reliability. Powerful and user-friendly, the 7300 Sequence Detection Software includes:
• Simple plate setup wizard is streamlined for each application – option to setup before or after the completing the run
• Correct plate-setup mistakes after the run without losing collected data.
• One-click graphical export to PowerPoint or JPG for top notch presentations
• Flexible system can be used as a plate reader or a regular thermal cycler if needed
• Real-time amplification monitoring and flexibility to add additional PCR cycles during a run
• Lamp-lifetime monitoring ensures highest possible instrument performance
For Research Use Only. Not for use in diagnostics procedures.
Spécification
Spécification
| Content And Storage | Store at Room Temperature |
| For Use With (Equipment) | 7300 System |
| Software Type | Automation Control |
| Product Type | Software Upgrade Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Operating System | Windows XP Pro (Service Pk. 2) |
| Quantity | 1 kit |
For Research Use Only. Not for use in diagnostic procedures.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.