Learn More
Ansell™ 70-225 Series Yellow Neptune™ Heavy Weight Gloves
Product Code. 12636921 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
10
Unit Size:
12 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12636921 10 12 pairs
12656921 8 12 pairs
12646921 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12636921 Supplier Ansell™ Supplier No. 70225100
Only null left
Add to basket
Add to basket
100% aramid fiber construction offers a very high cut resistance to weight ratio. Repels liquids, avoiding damage to the strands and extending glove life.
  • Made from high-tech Kevlar™ Aramid fibers para-aramid short fibers
  • Neptune coating allows glove fibers to repel liquids
  • Seamless construction to avoid irritation
  • Short fibers provide extra comfort
  • Terry-loop technique enchances heat insulation
  • Thick wall offers high shock absorption
  • Ambidextrous
  • Washable and reusable
TRUSTED_SUSTAINABILITY
Specifications
Back Material Neptune™
Certifications/Compliance Standards CATEGORY III EN 388 : 254X, EN 407, ANSI
Cuff Material Neptune™
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Heavyweight Gloves
Size 10
Glove Cuff Style Knit
Liner Material Dupont™ Kevlar™ aramid fibers
Lining Lined
Quantity 144 Pairs
Resistant To Very high cut resistance