missing translation for 'onlineSavingsMsg'
Learn More

7-Phenylheptanoic acid, 97%

Product Code. 11301797
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301797 1 g 1g
11311797 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301797 Supplier Thermo Scientific Alfa Aesar Supplier No. L05009.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40228-90-8 | C13H18O2 | 206.285 g/mol

7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40228-90-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00037801
InChI Key ZVSXKFNTWOIGJI-UHFFFAOYSA-N
Synonym benzeneheptanoic acid, phenylheptanoic acid, 7-phenyl-heptanoic acid, acmc-20alwn, 7-phenylheptanoic acid, xxc
PubChem CID 520987
IUPAC Name 7-phenylheptanoic acid
SMILES C1=CC=C(C=C1)CCCCCCC(=O)O

Specifications

Melting Point ∼20°C
Density 1.009
Boiling Point 147°C to 150°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 1 g
Beilstein 1957081
Solubility Information Insoluble in water.
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material 7-Phenylheptanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.