missing translation for 'onlineSavingsMsg'
Learn More

7-Phenyl-4-oxa-1-azabicyclo[4.2.0]octan-8-one, TRC

Product Code. 30423661
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423661 25 mg 25mg
30412644 50 mg 50mg
30417589 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423661 Supplier Toronto Research Chemicals Supplier No. TRCP33616325MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Phenyl-4-oxa-1-azabicyclo[4.2.0]octan-8-one
Molecular Formula C12 H13 N O2
Quantity 25 mg
InChI Formula InChI=1S/C12H13NO2/c14-12-11(9-4-2-1-3-5-9)10-8-15-7-6-13(10)12/h1-5,10-11H,6-8H2
SMILES O=C1C(C2COCCN12)c3ccccc3
IUPAC Name 7-phenyl-4-oxa-1-azabicyclo[4.2.0]octan-8-one
Molecular Weight (g/mol) 203.237
Formula Weight 203.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.