missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-p-Toluenesulfonylhydrazide Cholesterol-d7 3-Acetate, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | 7-p-Toluenesulfonylhydrazide Cholesterol-d7 3-Acetate |
| Molecular Formula | C36 D7 H47 N2 O4 S |
| Quantity | 1 mg |
| Synonym | 4-Methyl-benzenesulfonic Acid 2-[(3beta)-3-(Acetyloxy)cholest-5-en-7-ylidene]hydrazide-d7, p-Toluenesulfonic Acid (3beta-Hydroxycholest-5-en-7-ylidene)hydrazide-d7 Acetate |
| InChI Formula | InChI=1S/C36H54N2O4S/c1-23(2)9-8-10-25(4)30-15-16-31-34-32(18-20-36(30,31)7)35(6)19-17-28(42-26(5)39)21-27(35)22-33(34)37-38-43(40,41)29-13-11-24(3)12-14-29/h11-14,22-23,25,28,30-32,34,38H,8-10,15-21H2,1-7H3/b37-33-/t25-,28+,30-,31+,32+,34?,35+,36-/m1/s1/i1D3,2D3,23D |
| SMILES | [2H]C([2H])([2H])C([2H])(CCC[C@@H](C)[C@H]1CC[C@H]2C3[C@H](CC[C@]12C)[C@@]4(C)CC[C@@H](CC4=C/C/3=N/NS(=O)(=O)c5ccc(C)cc5)OC(=O)C)C([2H])([2H])[2H] |
| IUPAC Name | [(3S,7E,9S,10R,13R,14S,17R)-10,13-dimethyl-7-[(4-methylphenyl)sulfonylhydrazinylidene]-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-1,2,3,4,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate |
| Molecular Weight (g/mol) | 617.933 |
| Formula Weight | 617.424 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction