missing translation for 'onlineSavingsMsg'
Learn More

7-Oxabicyclo[2.2.1]heptane 99%, Thermo Scientific™

Product Code. 10467520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467520 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10467520 Supplier Thermo Scientific Acros Supplier No. 129590050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 279-49-2
CAS Min % 98.5
Color Colorless
CAS Max % 100.0
Density 0.9600g/mL
Quantity 5 g
Flash Point 12°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4470 to 1.4490
Assay Percent Range 99%
Specific Gravity 0.96
Molecular Formula C6H10O
MDL Number MFCD00082342
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
Boiling Point 119.0°C (713.0 mmHg)
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
SMILES C1CC2CCC1O2
IUPAC Name 7-oxabicyclo[2.2.1]heptane
Molecular Weight (g/mol) 98.15
PubChem CID 9234
Formula Weight 98.14
Percent Purity 99%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 7-Oxabicyclo[2.2.1]heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.