missing translation for 'onlineSavingsMsg'
Learn More

7-O-(Triethylsilyl)-2’-O-tert-butyl(dimethyl)silyl 2-Debenzoyl Paclitaxel 2-Pentanoate, TRC

Product Code. 30456371
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456371 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456371 Supplier Toronto Research Chemicals Supplier No. TRCT77784010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-O-(Triethylsilyl)-2'-O-tert-butyl(dimethyl)silyl 2-Debenzoyl Paclitaxel 2-Pentanoate
CAS 1055033-93-6
Molecular Formula C57H83NO14Si2
Quantity 10 mg
Synonym (alphaR,betaS)-beta-(Benzoylamino)-alpha-tert-butyl(dimethyl)silyloxy-benzenepropanoic Acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-Bis(acetyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4-triethylsilyloxy-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-12-[(1-oxopentyl)oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester
InChI Formula InChI=1S/C57H83NO14Si2/c1-16-20-31-43(61)69-50-48-55(13,41(71-74(17-2,18-3)19-4)32-42-56(48,34-66-42)70-37(7)60)49(62)46(67-36(6)59)44-35(5)40(33-57(50,65)54(44,11)12)68-52(64)47(72-73(14,15)53(8,9)10)45(38-27-23-21-24-28-38)58-51(63)39-29-25-22-26-30-39/h21-30,40-42,45-48,50,65H,16-20,31-34H2,1-15H3,(H,58,63)/t40-,41-,42+,45-,46+,47+,48-,50-,55+,56-,57+/m0/s1
SMILES CCCCC(=O)O[C@H]1[C@H]2[C@@](C)([C@H](C[C@H]3OC[C@@]23OC(=O)C)O[Si](CC)(CC)CC)C(=O)[C@H](OC(=O)C)C4=C(C)[C@H](C[C@]1(O)C4(C)C)OC(=O)[C@H](O[Si](C)(C)C(C)(C)C)[C@@H](NC(=O)c5ccccc5)c6ccccc6
Molecular Weight (g/mol) 1062.44
Formula Weight 1061.5352
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.