missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Product Code. 15497518
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497518 1 g 1g
15407528 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497518 Supplier Thermo Scientific Alfa Aesar Supplier No. H61032.03
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 120711-81-1 | Molecular Formula: C8H8N2O3 | Molecular Weight: 180.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120711-81-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD11603433
InChI Key YKCFDUNYLMTXFC-UHFFFAOYSA-N
Synonym 7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
PubChem CID 18416151
IUPAC Name 7-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Solubility Information Sparingly soluble in water (0.76g/L at 25°C).
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 7-Nitro-3,4-dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.