missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97+%

Product Code. 15449775
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449775 1 g 1g
15469775 250 mg 250mg
15459775 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449775 Supplier Thermo Scientific Alfa Aesar Supplier No. H52389.03
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 99365-69-2 | C9H11ClN2O2 | 214.649 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99365-69-2
Molecular Formula C9H11ClN2O2
Molecular Weight (g/mol) 214.649
MDL Number MFCD07371494
InChI Key KGIXLJRTYZOUCW-UHFFFAOYSA-N
Synonym 7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-7-nitroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 1,2,3,4-tetrahydro-7-nitro-, monohydrochloride, ksc683k9f, 7-nitro-1,2,3,4-tetrahydroisoquinoline monohydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 13521670
IUPAC Name 7-nitro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)[N+](=O)[O-].Cl

Specifications

Quantity 1 g
Formula Weight 214.65
Percent Purity ≥97%
Chemical Name or Material 7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.