missing translation for 'onlineSavingsMsg'
Learn More

7-Methylxanthine, MedChemExpress

Product Code. 16445457
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16445457 Supplier Medchem Express Supplier No. HYW017163
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7-Methylxanthine, a methyl derivative of xanthine, is one of the purine components in urinary calculi.

7-Methylxanthine, a methyl derivative of xanthine, is one of the purine components in urinary calculi.

Specifications

Chemical Name or Material 7-Methylxanthine
CAS 552-62-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H6N4O2
Quantity 50 mg
Solubility Information DMSO : 5 mg/mL (30.10 mM; Need ultrasonic)
SMILES CN1C=NC2=C1C(=O)NC(=O)N2
Molecular Weight (g/mol) 166.14
Formula Weight 166.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.