missing translation for 'onlineSavingsMsg'
Learn More

7-Methylindole-3-acetic Acid, TRC

Product Code. 30471010
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471010 50 mg 50mg
30483394 5 mg 5mg
30473923 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471010 Supplier Toronto Research Chemicals Supplier No. TRCM22387050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5435-36-9
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
IUPAC Name 2-(7-methyl-1H-indol-3-yl)acetic acid
SMILES Cc1cccc2c(CC(=O)O)c[nH]c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11NO2/c1-7-3-2-4-9-8(5-10(13)14)6-12-11(7)9/h2-4,6,12H,5H2,1H3,(H,13,14)
Formula Weight 189.079
Chemical Name or Material 7-Methylindole-3-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.