missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl Xanthine, TRC

Product Code. 30473317
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473317 Supplier Toronto Research Chemicals Supplier No. TRCM33858050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Methylxanthine
CAS 552-62-5
Molecular Formula C6 H6 N4 O2
Quantity 50 mg
Synonym 7-Methylxanthine, 1H-Purine-2,6-dione, 3,7-dihydro-7-methyl- (9CI, ACI), 3,7-Dihydro-7-methyl-1H-purine-2,6-dione (ACI), Heteroxanthine (6CI, 7CI, 8CI), 7-Methylxanthine, NSC 7861
InChI Formula InChI=1S/C6H6N4O2/c1-10-2-7-4-3(10)5(11)9-6(12)8-4/h2H,1H3,(H2,8,9,11,12)
SMILES Cn1cnc2NC(=O)NC(=O)c12
IUPAC Name 7-methyl-3H-purine-2,6-dione
Molecular Weight (g/mol) 166.14
Formula Weight 166.0491
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.