missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl lumazine

Product Code. 15476349
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476349 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476349 Supplier Thermo Scientific Alfa Aesar Supplier No. J64389.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13401-38-2 | C7H6N4O2 | 178.15 g/mol

A methyl-substituted imidazo[1,2-a]pyrazine derivative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A methyl-substituted imidazo[1,2-a]pyrazine derivative with anti-inflammatory, analgesic and ulcerogenic activities.
TRUSTED_SUSTAINABILITY

Specifications

CAS 13401-38-2
Molecular Formula C7H6N4O2
MDL Number MFCD19688352
Quantity 25 mg
Synonym 7-methyl lumazine, 7-methyllumazine, 7-methyllumizine, lumazine, 7-methyl, 7-methylpteridine-2,4 1h,3h-dione, 7-methylpteridine-2,4-diol, 2,4 1h,3h-pteridinedione, 7-methyl, 7-methyl-1,3-dihydropteridine-2,4-dione, 7-methyl-pteridine-2,4-diol, methyllumazin isomerengemisch
InChI Key WIXGFZWEUAUYGB-UHFFFAOYSA-N
SMILES CC1=CN=C2C(=O)NC(=O)NC2=N1
IUPAC Name 7-methyl-1H-pteridine-2,4-dione
Molecular Weight (g/mol) 178.15
PubChem CID 5374589
Formula Weight 178.15
Physical Form Powder
Chemical Name or Material 7-Methyl lumazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.