missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylic acid tert-butyl ester, Thermo Scientific™

Product Code. 10882421
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10882421

Brand: Thermo Scientific Maybridge MAY00279DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 243641-37-4
Packaging Amber Glass Bottle
Molecular Formula C14H20N2O2
Quantity 1 g
Synonym 7-methyl-3,4-dihydro-2h-1,8 naphthyridine-1-carboxylic acid tert-butyl ester, tert-butyl 7-methyl-3,4-dihydro-1,8-naphthyridine-1 2h-carboxylate, 1-boc-7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 1-boc-7-methyl-3,4-dihydro-2h-1,8 naphthyridine, 7-methyl-3,4-dihydro-2h-1,8-naphthyridine-1-carboxylic acid tert-butyl ester, 2-methyl-8-tert-butoxycarbonyl-5,6,7,8-tetrahydro-1,8-naphthyridine, tert-butyl 7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridinecarboxylate, tert-butyl 7-methyl-3,4-dihydro-2h-1,8-naphthyridine-1-carboxylate
InChI Key CHWMFCCGEWQABK-UHFFFAOYSA-N
SMILES CC1=NC2=C(CCCN2C(=O)OC(C)(C)C)C=C1
IUPAC Name tert-butyl 7-methyl-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate
Molecular Weight (g/mol) 248.326
PubChem CID 17750173
Formula Weight 248.32
Percent Purity 95%
Chemical Name or Material 7-Methyl-3, 4-dihydro-2H-1, 8-naphthyridine-1-carboxylic acid tert-butyl ester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.