Learn More
7-Methoxycoumarin-4-acetic acid
Description
7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.
Solubility
Soluble in acetone and dimethyl sulfoxide.
Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 193°C |
| Color | Green |
| Flash Point | 87°C (189°F) |
| Quantity | 1 g |
| Solubility Information | Soluble in acetone and dimethyl sulfoxide. |
| Formula Weight | 234.21 |
| Physical Form | Crystalline Solid |
| Chemical Name or Material | 7-Methoxycoumarin-4-acetic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.