missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-6-(3-morpholin-4-ylpropoxy)quinazolin-4(3H)-one, TRC

Product Code. 30138499
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138499 250 mg 250mg
30136558 100 mg 100mg
30006433 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138499 Supplier Toronto Research Chemicals Supplier No. TRCM264670250MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199327-61-2
Molecular Formula C16 H21 N3 O4
Molecular Weight (g/mol) 319.36
Synonym 4(3H)-Quinazolinone, 7-methoxy-6-[3-(4-morpholinyl)propoxy]-, 4(1H)-Quinazolinone, 7-methoxy-6-[3-(4-morpholinyl)propoxy]- (9CI), 7-Methoxy-6-[3-(4-morpholinyl)propoxy]-4(3H)-quinazolinone, 7-Methoxy-6-(3-(morpholin-4-yl)propoxy)-3H-quinazolin-4-one, 7-Methoxy-6-(3-morpholinopropoxy)-3,4-dihydroquinazolin-4-one, 7-Methoxy-6-[3-(4-morpholinyl)propoxy]quinazolin-4(3H)-one, Gefitinib Imp. A (EP)
IUPAC Name 7-methoxy-6-(3-morpholin-4-ylpropoxy)-3H-quinazolin-4-one
SMILES COc1cc2N=CNC(=O)c2cc1OCCCN3CCOCC3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H21N3O4/c1-21-14-10-13-12(16(20)18-11-17-13)9-15(14)23-6-2-3-19-4-7-22-8-5-19/h9-11H,2-8H2,1H3,(H,17,18,20)
Formula Weight 319.1532
Chemical Name or Material 7-Methoxy-6-(3-morpholin-4-ylpropoxy)quinazolin-4(3H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.