missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-4-methylcoumarin, 99%, Thermo Scientific™

Product Code. 11429363
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429363 1 g 1g
11439363 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429363 Supplier Thermo Scientific Alfa Aesar Supplier No. A11785.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A coumarin derivative and fluorescent label.

Solubility
Soluble in Methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2555-28-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00009773
InChI Key UDFPKNSWSYBIHO-UHFFFAOYSA-N
Synonym 7-methoxy-4-methylcoumarin, 7-methoxy-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-methoxy-4-methyl, 4-methylherniarin, hymecromone methyl ether, herniarin, 4-methyl, 7-methoxy-4-methyl-chromen-2-one, 4-methyl-7-methoxycoumarin, coumarin,4-methyl-7-methoxy, coumarin, 7-methoxy-4-methyl
PubChem CID 390807
IUPAC Name 7-methoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC

Specifications

Melting Point 159°C to 162°C
Quantity 1 g
Beilstein 156900
Solubility Information Soluble in Methanol. Insoluble in water.
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material 7-Methoxy-4-methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.