missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2-naphthol, 97%, Thermo Scientific Chemicals

Catalog Number p-4089698
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5060-82-2
C11H10O2
174.20
MFCD00075494
UNFNRIIETORURP-UHFFFAOYSA-N
7-methoxy-2-naphthol, 2-naphthalenol, 7-methoxy, 7-methoxy-2-naphthalenol, pubchem16354, 7-methoxy-beta-naphthol, acmc-1bnbv, 7-methoxy-naphthalen-2-ol, 7-methyloxy-2-naphthalenol, 2-hydroxy-7-methoxynaphthalene
853173
7-methoxynaphthalen-2-ol
COC1=CC=C2C=CC(O)=CC2=C1
This item is not returnable. View return policy
5060-82-2
C11H10O2
174.20
MFCD00075494
UNFNRIIETORURP-UHFFFAOYSA-N
7-methoxy-2-naphthol, 2-naphthalenol, 7-methoxy, 7-methoxy-2-naphthalenol, pubchem16354, 7-methoxy-beta-naphthol, acmc-1bnbv, 7-methoxy-naphthalen-2-ol, 7-methyloxy-2-naphthalenol, 2-hydroxy-7-methoxynaphthalene
853173
7-methoxynaphthalen-2-ol
COC1=CC=C2C=CC(O)=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 116.0°C to 119.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 174.2
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 7-Methoxy-2-naphthol
Show More Show Less
compliance-icons
Product Identifier
  • 7-Methoxy-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'