missing translation for 'onlineSavingsMsg'
Learn More

7-Keto Dehydro Epiandrosterone, TRC

Product Code. 30389226
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389226 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389226 Supplier Toronto Research Chemicals Supplier No. TRCK18770050MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Keto Dehydroisoandrosterone (7-Keto DHEA)
CAS 566-19-8
Purity Grade Notes HPLC
Molecular Formula C19 H26 O3
Quantity 50 mg
Synonym 7-Ketoprasterone, Androst-5-ene-7,17-dione, 3-hydroxy-, (3β)-, Androst-5-ene-7,17-dione, 3β-hydroxy- (8CI), (3β)-3-Hydroxyandrost-5-ene-7,17-dione, 3β-Hydroxy-5-androstene-7,17-dione, 5-Androsten-3β-ol-7,17-dione, 7-Keto-DHEA, 7-Ketodehydroepiandrosterone, 7-Oxo-DHEA, 7-Oxodehydroepiandrosterone, 7-keto-DHEA
InChI Formula InChI=1S/C19H26O3/c1-18-7-5-12(20)9-11(18)10-15(21)17-13-3-4-16(22)19(13,2)8-6-14(17)18/h10,12-14,17,20H,3-9H2,1-2H3/t12-,13-,14-,17-,18-,19-/m0/s1
SMILES C[C@]12CC[C@H]3[C@H]([C@@H]1CCC2=O)C(=O)C=C4C[C@@H](O)CC[C@]34C
IUPAC Name (3S,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-2,3,4,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-7,17-dione
Molecular Weight (g/mol) 302.41
Formula Weight 302.1882
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.