missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyquinoline, TRC

Product Code. 30404347
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 30404347

Brand: Toronto Research Chemicals TRCH9542082.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxyquinoline
CAS 580-20-1
Molecular Formula C9 H7 N O
Quantity 2.5 g
Synonym 7-Hydroxyquinoline
InChI Formula InChI=1S/C9H7NO/c11-8-4-3-7-2-1-5-10-9(7)6-8/h1-6,11H
SMILES Oc1ccc2cccnc2c1
IUPAC Name quinolin-7-ol
Molecular Weight (g/mol) 145.158
Formula Weight 145.053
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.