missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Hydroxyquinoline, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 7-Hydroxyquinoline |
| CAS | 580-20-1 |
| Molecular Formula | C9 H7 N O |
| Quantity | 10 g |
| Synonym | 7-Hydroxyquinoline |
| InChI Formula | InChI=1S/C9H7NO/c11-8-4-3-7-2-1-5-10-9(7)6-8/h1-6,11H |
| SMILES | Oc1ccc2cccnc2c1 |
| IUPAC Name | quinolin-7-ol |
| Molecular Weight (g/mol) | 145.158 |
| Formula Weight | 145.053 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction