missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Ondansetron, TRC

Product Code. 30390529
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390529 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390529 Supplier Toronto Research Chemicals Supplier No. TRCH9511501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxy Ondansetron
CAS 126702-17-8
Molecular Formula C18 H19 N3 O2
Quantity 1 mg
Synonym 4H-Carbazol-4-one, 1,2,3,9-tetrahydro-7-hydroxy-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-, 1,2,3,9-Tetrahydro-7-hydroxy-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one, 7-Hydroxyondansetron, GR 63418
InChI Formula InChI=1S/C18H19N3O2/c1-11-19-7-8-21(11)10-12-3-6-15-17(18(12)23)14-5-4-13(22)9-16(14)20(15)2/h4-5,7-9,12,22H,3,6,10H2,1-2H3
SMILES Cc1nccn1CC2CCc3c(C2=O)c4ccc(O)cc4n3C
IUPAC Name 7-hydroxy-9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one
Molecular Weight (g/mol) 309.362
Formula Weight 309.148
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.