missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Loxapine, TRC

Product Code. 30390857
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390857 2.5 mg 2.5mg
30401078 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390857 Supplier Toronto Research Chemicals Supplier No. TRCH9437902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37081-75-7
Molecular Formula C18H18ClN3O2
Molecular Weight (g/mol) 343.81
Synonym 2-Chloro-11-(4-methyl-1-piperazinyl)-dibenz[b,f][1,4]oxazepin-7-ol, 2-Chloro-7-hydroxy-11-(4-methyl-1-piperazinyl)dibenz[b,f][1,4]oxazepine, 2-Chloro-7-hydroxy-11-(4-methyl-1-piperazinyl)dibenz[b,f]oxazepine
IUPAC Name 8-chloro-6-(4-methylpiperazin-1-yl)benzo[b][1,4]benzoxazepin-2-ol
SMILES CN1CCN(CC1)C2=Nc3ccc(O)cc3Oc4ccc(Cl)cc24

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C18H18ClN3O2/c1-21-6-8-22(9-7-21)18-14-10-12(19)2-5-16(14)24-17-11-13(23)3-4-15(17)20-18/h2-5,10-11,23H,6-9H2,1H3
Formula Weight 343.1088
Chemical Name or Material 7-Hydroxy Loxapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.