missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy DAMPA, TRC

Product Code. 30389305
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389305 10 mg 10mg
30406577 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389305 Supplier Toronto Research Chemicals Supplier No. TRCH93150010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34698-85-6
Molecular Formula C15 H15 N7 O3
Molecular Weight (g/mol) 341.32
Synonym Benzoic acid, 4-[[(2,4-diamino-7,8-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]-, Benzoic acid, 4-[[(2,4-diamino-1,7-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]- (9CI), 4-[[(2,4-Diamino-7,8-dihydro-7-oxo-6-pteridinyl)methyl]methylamino]benzoic acid, 4-Amino-4-deoxy-7-hydroxy-N10-methylpteroic acid
IUPAC Name 4-[(2,4-diamino-7-hydroxypteridin-6-yl)methyl-methylamino]benzoic acid
SMILES CN(Cc1nc2c(N)nc(N)nc2nc1O)c3ccc(cc3)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H15N7O3/c1-22(8-4-2-7(3-5-8)14(24)25)6-9-13(23)20-12-10(18-9)11(16)19-15(17)21-12/h2-5H,6H2,1H3,(H,24,25)(H5,16,17,19,20,21,23)
Formula Weight 341.1236
Chemical Name or Material 7-Hydroxy DAMPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.