missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin, TRC

Product Code. 30136484
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136484 1 g 1g
30006999 2.5 g 2.5g
17935015 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136484 Supplier Toronto Research Chemicals Supplier No. TRCH9248751G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-35-6
Molecular Formula C9 H6 O3
Molecular Weight (g/mol) 162.14
Synonym 2H-1-Benzopyran-2-one, 7-hydroxy-, Coumarin, 7-hydroxy- (7CI,8CI), 7-Hydroxy-2H-1-benzopyran-2-one, 7-Hydroxy-2-chromenone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxycoumarin, 7-Hydroxylcoumarin, 7-Oxycoumarin, Hydrangin, Hydrangine, NSC 19790, Skimmetin, Skimmetine, Umbelliferon, Umbelliferone
IUPAC Name 7-hydroxychromen-2-one
SMILES Oc1ccc2C=CC(=O)Oc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H
Formula Weight 162.0317
Percent Purity >95%
Chemical Name or Material 7-Hydroxycoumarin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.