missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin-d5 Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30392876
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392876 1 mg
30391846 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392876 Supplier Toronto Research Chemicals Supplier No. TRCH9248821MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H8D5NaO9
Molecular Weight (g/mol) 365.28
Synonym 2-Oxo-2H-1-benzopyran-7-yl-d7 β-D-Glucopyranosiduronic Acid Monosodium Salt
IUPAC Name sodium (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((2-oxo-2H-chromen-7-yl-3,4,5,6,8-d5)oxy)tetrahydro-2H-pyran-2-carboxylate
SMILES O=C1OC2=C([2H])C(O[C@@H]([C@@H]3O)O[C@H](C(O[Na])=O)[C@@H](O)[C@@H]3O)=C([2H])C([2H])=C2C([2H])=C1[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H14O9.Na/c16-9-4-2-6-1-3-7(5-8(6)23-9)22-15-12(19)10(17)11(18)13(24-15)14(20)21;/h1-5,10-13,15,17-19H,(H,20,21);/q;+1/p-1/t10-,11-,12+,13-,15+;/m0./s1/i1D,2D,3D,4D,5D;
Formula Weight 365.08
Chemical Name or Material 7-Hydroxy Coumarin-d5 b-D-Glucuronide Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.