missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin-13C6, TRC

Product Code. 30400608
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400608 10 mg
30403688 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400608 Supplier Toronto Research Chemicals Supplier No. TRCH92487810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189992-05-9
Molecular Formula C313C6H6O3
Molecular Weight (g/mol) 168.1
Synonym 7-Hydroxy-2H-1-benzopyran-2-one-4a,5,6,7,8,8a-13C6, 7-Hydroxy-2H-chromen-2-one-4a,5,6,7,8,8a-13C6, 7-Hydroxy coumarin-13C6, 7-Hydroxycoumarin-13C6, 7-Hydroxycoumarin-4a,5,6,7,8,8a-13C6, Umbelliferone-13C6, Umbelliferone-4a,5,6,7,8,8a-13C6
IUPAC Name 7-hydroxychromen-2-one
SMILES O[13c]1[13cH][13cH][13c]2C=CC(=O)O[13c]2[13cH]1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H/i1+1,3+1,5+1,6+1,7+1,8+1
Formula Weight 168.0518
Chemical Name or Material 7-Hydroxy Coumarin-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.