missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Amoxapine, TRC

Product Code. 17984515
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984515 1 mg 1mg
30132405 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984515 Supplier Toronto Research Chemicals Supplier No. TRCH7978001MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37081-76-8
Molecular Weight (g/mol) 329.78
IUPAC Name 8-chloro-6-piperazin-1-ylbenzo[b][1,4]benzoxazepin-2-ol
SMILES Oc1ccc2N=C(N3CCNCC3)c4cc(Cl)ccc4Oc2c1

Specifications

Molecular Formula (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Quantity 1 mg
InChI Formula InChI=1S/C17H16ClN3O2/c18-11-1-4-15-13(9-11)17(21-7-5-19-6-8-21)20-14-3-2-12(22)10-16(14)23-15/h1-4,9-10,19,22H,5-8H2
Formula Weight 329.0931
Chemical Name or Material 7-Hydroxy Amoxapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.