missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline, 98%, Thermo Scientific™

Product Code. 10119931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119931 1 g Glass bottle 1g
10540892 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10119931 Supplier Thermo Scientific Acros Supplier No. 277820010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4602-73-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00143517
InChI Key UQBWYWCIBNWMPI-UHFFFAOYSA-N
Synonym 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
PubChem CID 363043
IUPAC Name 6-methoxy-3,4-dihydroisoquinolin-7-ol
SMILES COC1=C(C=C2C=NCCC2=C1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 191°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in chloroform,ethanol,methanol and hot,ethyl acetate
Formula Weight 177.2
Percent Purity 98%
Chemical Name or Material 7-Hydroxy-6-methoxy-3, 4-dihydroisoquinoline, 98%
compliance-icons
Product Identifier
  • 7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.