missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-5-methoxy-4-methyl-6-[2-(tetrahydro-2-methyl-5-oxo-2-furanyl)ethyl]-1(3H)-isobenzofuranone, TRC

Product Code. 30396447
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
5mg
Product Code. Quantity unitSize
30396447 5 mg 5mg
30403176 50 mg 50mg
30395214 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30396447

Brand: Toronto Research Chemicals TRCH8050855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26675-76-3
Formula Weight 320.126
InChI Formula InChI=1S/C17H20O6/c1-9-11-8-22-16(20)13(11)14(19)10(15(9)21-3)4-6-17(2)7-5-12(18)23-17/h19H,4-8H2,1-3H3
IUPAC Name 7-hydroxy-5-methoxy-4-methyl-6-[2-(2-methyl-5-oxooxolan-2-yl)ethyl]-3H-2-benzofuran-1-one
Molecular Formula C17 H20 O6
Molecular Weight (g/mol) 320.34
SMILES COc1c(C)c2COC(=O)c2c(O)c1CCC3(C)CCC(=O)O3
Synonym 1(3H)-Isobenzofuranone, 7-hydroxy-5-methoxy-4-methyl-6-[2-(tetrahydro-2-methyl-5-oxo-2-furanyl)ethyl]- (9CI), Phthalide, 7-hydroxy-5-methoxy-4-methyl-6-[2-(tetrahydro-2-methyl-5-oxo-2-furyl)ethyl]- (8CI), 7-Hydroxy-5-methoxy-4-methyl-6-[2-[(2RS)-2-methyl-5-oxotetrahydrofuran-2-yl]ethyl]isobenzofuran-1(3H)-one, Mycophenolate Mofetil Imp. H (EP), Mycophenolate Sodium Imp. A (EP), Mycophenolic Acid Lactone
Chemical Name or Material 7-Hydroxy-5-methoxy-4-methyl-6-[2-[(2RS)-2-methyl-5-oxotetrahydrofuran-2-yl]ethyl]isobenzofuran-1(3H)-one

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.