missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt, TRC

Product Code. 30393719
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393719 10 mg 10mg
30403231 1 mg 1mg
30408569 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393719 Supplier Toronto Research Chemicals Supplier No. TRCH92480510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt
Analyte or Component Names 7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt
Shipping Condition Room Temperature
Molecular Formula C16 H16 O9 . C6 H13 N
Synonym (2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((5-methyl-7-oxo-7,8-dihydronaphthalen-2-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid cyclohexamine
InChI Formula InChI=1S/C16H16O9.C6H13N/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;7-6-4-2-1-3-5-6/h2-5,11-14,16,18-20H,1H3,(H,21,22);6H,1-5,7H2/t11-,12-,13+,14-,16+;/m0./s1
Solution Type Neat
SMILES CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12.NC4CCCCC4
IUPAC Name cyclohexanamine;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 451.467
Quantity 10 mg
Formula Weight 451.184
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.