missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-3,7-dimethyloctanal, MP Biomedicals

Product Code. 11468795 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468795 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468795 Supplier MP Biomedicals Supplier No. 0522493910
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107-75-5
Molecular Formula C10H20O2
Quantity 10 g
Synonym hydroxycitronellal, 7-hydroxycitronellal, citronellal hydrate, 3,7-dimethyl-7-hydroxyoctanal, phixia, cyclalia, cyclosia, laurine, fixol, lilyl aldehyde
InChI Key WPFVBOQKRVRMJB-UHFFFAOYSA-N
SMILES CC(CCCC(C)(C)O)CC=O
IUPAC Name 7-hydroxy-3,7-dimethyloctanal
Molecular Weight (g/mol) 172.268
PubChem CID 7888
ChEBI CHEBI:53459
Formula Weight 172
Chemical Name or Material 7-Hydroxy-3, 7-Dimethyl Octanal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.