missing translation for 'onlineSavingsMsg'
Learn More

7-[(Hydroxy-2-thienyl-3-thienylacetyl)oxy]-9,9-dimethyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide-d6, TRC

Product Code. 30405540
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405540 100 mg 100mg
30398057 25 mg 25mg
30390814 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30405540 Supplier Toronto Research Chemicals Supplier No. TRCH962027100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-[(Hydroxy-2-thienyl-3-thienylacetyl)oxy]-9,9-dimethyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide-d6
Molecular Formula C19 D6 H16 N O4 S2
Quantity 100 mg
InChI Formula InChI=1S/C19H22NO4S2/c1-20(2)13-8-12(9-14(20)17-16(13)24-17)23-18(21)19(22,11-5-7-25-10-11)15-4-3-6-26-15/h3-7,10,12-14,16-17,22H,8-9H2,1-2H3/q+1/t12-,13-,14+,16-,17?,19?/m1/s1/i1D3,2D3
SMILES [2H]C([2H])([2H])[N+]1([C@@H]2C[C@H](C[C@H]1C3O[C@H]23)OC(=O)C(O)(c4ccsc4)c5cccs5)C([2H])([2H])[2H]
IUPAC Name [(1S,4R,5R)-9,9-bis(trideuteriomethyl)-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonan-7-yl] 2-hydroxy-2-thiophen-2-yl-2-thiophen-3-ylacetate
Molecular Weight (g/mol) 398.549
Formula Weight 398.136
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.