missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1,4-benzodioxan-6-carboxylic Acid, TRC

Product Code. 30389567
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389567 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389567 Supplier Toronto Research Chemicals Supplier No. TRCH82919025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxy-1,4-benzodioxan-6-carboxylic Acid
CAS 197584-99-9
Molecular Formula C9H8O5
Quantity 25 mg
Synonym 2,3-Dihydro-7-hydroxy-1,4-benzodioxin-6-carboxylic Acid
InChI Formula InChI=1S/C9H8O5/c10-6-4-8-7(13-1-2-14-8)3-5(6)9(11)12/h3-4,10H,1-2H2,(H,11,12)
SMILES OC(=O)c1cc2OCCOc2cc1O
IUPAC Name 6-hydroxy-2,3-dihydro-1,4-benzodioxine-7-carboxylic acid
Molecular Weight (g/mol) 196.16
Formula Weight 196.0372
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.