missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1,2,3,4-tetrahydroquinoline, 96%

Product Code. 15587519 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15587519 1 g 1g
15597519 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15587519 Supplier Thermo Scientific Acros Supplier No. 458710010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58196-33-1 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58196-33-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
InChI Key HJJRGZMJZDSMDB-UHFFFAOYSA-N
Synonym 7-hydroxy-1,2,3,4-tetrahydroquinoline, 7-quinolinol, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-7-quinolinol, 1,2,3,4-tetrahydro-quinolin-7-ol, 1,2,3,4-tetrahydrochinolin-7-ol, pubchem12787, 7-hydroxy-3,4-dihydro-2h-quinoline, 1,2,3,4-tetrahydro-7-hydroxyquinoline, 7-hydroxy-1,2,3,4-tetrahydro-quinolin, 7-hydroxy-1,2,3,4-tetrahydro quinolinoe
PubChem CID 93980
IUPAC Name 1,2,3,4-tetrahydroquinolin-7-ol
SMILES C1CC2=C(C=C(C=C2)O)NC1

Specifications

Melting Point 91.0°C to 93.5°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 149.19
Percent Purity 96%
Chemical Name or Material 7-Hydroxy-1,2,3,4-tetrahydroquinoline
compliance-icons
Product Identifier
  • 7-Hydroxy-1,2,3,4-tetrahydroquinoline, 96%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.