missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoroisatin, 97%

Product Code. 11326969
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326969 1 g 1g
11336969 5 g 5g
11346969 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326969 Supplier Thermo Scientific Alfa Aesar Supplier No. H26827.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 317-20-4 | C8H4FNO2 | 165.123 g/mol

7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

Solubility
Sparingly Soluble in water (0.022 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317-20-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
MDL Number MFCD01569508
InChI Key HGBGVEOXPHGSOS-UHFFFAOYSA-N
Synonym 7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion
PubChem CID 586418
IUPAC Name 7-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)F)NC(=O)C2=O

Specifications

Melting Point 193°C to 195°C
Quantity 1 g
Solubility Information Sparingly Soluble in water (0.022g/L)
Formula Weight 165.12
Percent Purity 97%
Chemical Name or Material 7-Fluoroisatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.