missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro Hydroxychloroquine, TRC

Product Code. 30406399
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406399 100 mg 100mg
30397865 250 mg -
30392386 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406399 Supplier Toronto Research Chemicals Supplier No. TRCF588890100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 478784-65-5
Molecular Formula C18H26FN3O
Molecular Weight (g/mol) 319.42
Synonym 2-[Ethyl[4-[(7-fluoro-4-quinolinyl)amino]pentyl]amino]ethanol
SMILES CCN(CCO)CCCC(NC1=C2C=CC(F)=CC2=NC=C1)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H26FN3O/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21)
Chemical Name or Material 7-Fluoro Hydroxychloroquine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.