missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-6-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Product Code. 15416266
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416266 250 mg 250mg
15447588 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416266 Supplier Thermo Scientific Alfa Aesar Supplier No. H61092.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in medicine.

Solubility
Slightly soluble in water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103361-67-7
Molecular Formula C8H5FN2O4
Molecular Weight (g/mol) 212.136
MDL Number MFCD22056242
InChI Key MNUBUFBVOBPANC-UHFFFAOYSA-N
Synonym 2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro, 7-fluoro-6-nitro-2h-1,4-benzoxazin-3 4h-one, 7-fluoro-6-nitro-2,4-dihydro-1,4-benzoxazin-3-one, acmc-20m67v, 7-fluoro-6-nitro-4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazin-3 4h-one,7-fluoro-6-nitro, 7-fluoro-3,4-dihydro-3-oxo-6-nitro-2h-1,4-benzoxazine
PubChem CID 13731297
IUPAC Name 7-fluoro-6-nitro-4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=CC(=C(C=C2O1)F)[N+](=O)[O-]

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 212.14
Percent Purity 97%
Chemical Name or Material 7-Fluoro-6-nitro-2H-1,4-benzoxazin-3(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.