missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole, 95%, Thermo Scientific™

Product Code. 11392959
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
11392959 100 mg 100mg
11302969 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 11392959

Brand: Thermo Scientific Alfa Aesar H27780.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177858-80-9
Molecular Formula C11H11FN2
Molecular Weight (g/mol) 190.221
MDL Number MFCD09264527
InChI Key OTGGOPSBNVEBDA-UHFFFAOYSA-N
Synonym 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro
PubChem CID 15077550
IUPAC Name 7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES C1CNCC2=C1C3=C(N2)C=C(C=C3)F

Specifications

Melting Point 216°C to 224°C
Quantity 100 mg
Formula Weight 190.22
Percent Purity 95%
Chemical Name or Material 7-Fluoro-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.