missing translation for 'onlineSavingsMsg'
Learn More

7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidino)carbonyloxycamptothecin, TRC

Product Code. 30383586
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383586 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383586 Supplier Toronto Research Chemicals Supplier No. TRCE8989001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Ethyl-10-(4-N-aminopentanoic acid)-1-piperidino)carbonyloxycamptothecin
CAS 181467-56-1
Molecular Formula C33 H38 N4 O8
Quantity 1 mg
InChI Formula InChI=1S/C33H38N4O8/c1-3-21-22-15-20(45-32(42)36-13-10-19(11-14-36)34-12-6-5-7-28(38)39)8-9-26(22)35-29-23(21)17-37-27(29)16-25-24(30(37)40)18-44-31(41)33(25,43)4-2/h8-9,15-16,19,34,43H,3-7,10-14,17-18H2,1-2H3,(H,38,39)/t33-/m0/s1
SMILES CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(CC6)NCCCCC(=O)O)cc15)[C@@](O)(CC)C(=O)OC4
Molecular Weight (g/mol) 618.6768
Formula Weight 618.269
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.