missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 7-Ethoxyresorufin

Product Code. 18749606
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749606 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749606 Supplier R&D Systems™ Supplier No. 6204/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorometric CYP450 substrate

7-Ethoxyresorufin is a fluorometric substrate of cytochrome P450.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5725-91-7
Quantity 10 mg
Product Line Tocris
Target Cytochrome P450
Molecular Formula C14H11NO3
Formula Weight 241.24g/mol
Product Type Substrate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.