missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-epi-Paclitaxel, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
7-epi-Paclitaxel
Specifications
Specifications
| Chemical Name or Material | 7-Epitaxol |
| CAS | 105454-04-4 |
| Analyte or Component Names | 7-Epitaxol |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C47 H51 N O14 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 7-Epipaclitaxel, 7-Epitaxol, 7-epi-Paclitaxel, 7-epi-Taxol, Epitaxol, Paclitaxel Imp E (EP), Benzenepropanoic acid, beta-(benzoylamino)-alpha-hydroxy-, (2aR,4R,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (alphaR,betaS)-, Benzenepropanoic acid, beta-(benzoylamino)-alpha-hydroxy-, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aalpha,4alpha,4abeta,6beta,9alpha(alphaR*,betaS*),11alpha,12alpha,12aalpha,12balpha]]-, Paclitaxel Impurity E |
| InChI Formula | InChI=1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32+,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1 |
| SMILES | CC(=O)O[C@H]1C(=O)[C@]2(C)[C@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7)C(=C1C5(C)C)C |
| Molecular Weight (g/mol) | 853.91 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction