missing translation for 'onlineSavingsMsg'
Learn More

7-Epi-docetaxel, MedChemExpress

Product Code. 16487118
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487118 10 mg 10mg
16461629 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487118 Supplier Medchem Express Supplier No. HY16676
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7-Epi-10-oxo-docetaxel (Docetaxel Impurity C; 7-Epitaxotere) is a impurity of docetaxel.

7-Epi-10-oxo-docetaxel (Docetaxel Impurity C

Specifications

Chemical Name or Material 7-Epi-docetaxel
CAS 153381-68-1
Color White
Purity Grade Notes Research
Molecular Formula C43H53NO14
Quantity 10 mg
Synonym 4-epi-Docetaxel 7-Epidocetaxel 7-Epitaxotere
Solubility Information DMSO : 100 mg/mL (123.78 mM; Need ultrasonic)
SMILES CC(O[C@@]1([C@](C[C@H]2O)([H])OC1)[C@@]([C@@]2(C3=O)C)([H])[C@@H]([C@@](C4(C)C)(C[C@H](OC([C@H](O)[C@H](C5=CC=CC=C5)NC(OC(C)(C)C)=O)=O)C(C)=C4[C@H]3O)O)OC(C6=CC=CC=C6)=O)=O
Molecular Weight (g/mol) 807.88
Formula Weight 807.88
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.